missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pyridyl Trifluoromethanesulfonate 97.0+%, TCI America™

Catalog No. M19741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19741G 1 g
M19745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19741G Supplier TCI America Supplier No. M19741G
Only null left

Chemical Identifiers

CAS 179260-78-7
Molecular Formula C7H6F3NO3S
Molecular Weight (g/mol) 241.184
InChI Key AZUJRVRRUXHHDV-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 4-Methyl-2-pyridyl Ester
PubChem CID 10857619
IUPAC Name (4-methylpyridin-2-yl) trifluoromethanesulfonate
SMILES CC1=CC(=NC=C1)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 241.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pyridyl Trifluoromethanesulfonate
TSCA No
Recommended Storage Refrigerator