missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-phenylpyridine 97.0+%, TCI America™
$127.07 - $510.16
Chemical Identifiers
| CAS | 3475-21-6 |
|---|---|
| Molecular Formula | C12H11N |
| Molecular Weight (g/mol) | 169.23 |
| MDL Number | MFCD00087695 |
| InChI Key | WWMRJCUZPJJWBC-UHFFFAOYSA-N |
| Synonym | 2-Phenyl-4-picoline |
| PubChem CID | 77026 |
| IUPAC Name | 4-methyl-2-phenylpyridine |
| SMILES | CC1=CC=NC(=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
M2952200MG
|
TCI America
M2952200MG |
200 mg |
Each for $127.07
|
|
|||||
|
M29521G
|
TCI America
M29521G |
1 g |
Each for $510.16
|
|
|||||
Chemical Identifiers
| 3475-21-6 | |
| 169.23 | |
| WWMRJCUZPJJWBC-UHFFFAOYSA-N | |
| 77026 | |
| CC1=CC=NC(=C1)C1=CC=CC=C1 |
| C12H11N | |
| MFCD00087695 | |
| 2-Phenyl-4-picoline | |
| 4-methyl-2-phenylpyridine |
Specifications
| 3475-21-6 | |
| White-Yellow | |
| C12H11N | |
| 200 mg | |
| WWMRJCUZPJJWBC-UHFFFAOYSA-N | |
| 4-methyl-2-phenylpyridine | |
| 77026 | |
| ≥97.0% (GC,T) | |
| 4-Methyl-2-phenylpyridine |
| 49°C | |
| 115°C | |
| MFCD00087695 | |
| 2-Phenyl-4-picoline | |
| CC1=CC=NC(=C1)C1=CC=CC=C1 | |
| 169.23 | |
| 169.23 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No