missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenylimidazole 93.0+%, TCI America™

$89.48 - $89.48

Chemical Identifiers

CAS 827-43-0
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00047015
InChI Key TYOXIFXYEIILLY-UHFFFAOYSA-N
PubChem CID 70012
IUPAC Name 5-methyl-2-phenyl-1H-imidazole
SMILES CC1=CN=C(N1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M066925G
View Documents
TCI America
M066925G
25 g
Each for $89.48
Only null left
 
Description

Description

Specifications

Chemical Identifiers

827-43-0
158.20
TYOXIFXYEIILLY-UHFFFAOYSA-N
5-methyl-2-phenyl-1H-imidazole
C10H10N2
MFCD00047015
70012
CC1=CN=C(N1)C1=CC=CC=C1

Specifications

827-43-0
White-Yellow
MFCD00047015
4-methyl-2-phenyl-1h-imidazole, 4-methyl-2-phenylimidazole, 2-phenyl-4-methylimidazole, 1h-imidazole, 4-methyl-2-phenyl, 1h-imidazole, 5-methyl-2-phenyl, 5-methyl-2-phenylimidazole, methyl-2-phenylimidazole, acmc-209pq6, 1h-imidazole,5-methyl-2-phenyl
CC1=CN=C(N1)C1=CC=CC=C1
158.20
158.20
Crystalline Powder
181°C
C10H10N2
25 g
TYOXIFXYEIILLY-UHFFFAOYSA-N
5-methyl-2-phenyl-1H-imidazole
70012
≥93.0% (GC)
4-Methyl-2-phenylimidazole
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0411

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'