missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenylimidazole 93.0+%, TCI America™

Catalog No. M066925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M066925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M066925G Supplier TCI America Supplier No. M066925G
Only null left

Chemical Identifiers

CAS 827-43-0
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00047015
InChI Key TYOXIFXYEIILLY-UHFFFAOYSA-N
PubChem CID 70012
IUPAC Name 5-methyl-2-phenyl-1H-imidazole
SMILES CC1=CN=C(N1)C1=CC=CC=C1

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 25 g
Synonym 4-methyl-2-phenyl-1h-imidazole, 4-methyl-2-phenylimidazole, 2-phenyl-4-methylimidazole, 1h-imidazole, 4-methyl-2-phenyl, 1h-imidazole, 5-methyl-2-phenyl, 5-methyl-2-phenylimidazole, methyl-2-phenylimidazole, acmc-209pq6, 1h-imidazole,5-methyl-2-phenyl
Formula Weight 158.20
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-2-phenylimidazole
EINECSNumber (5)-0411
TSCA Yes