missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Methyl-2-phenyl-5-pyrimidinyl)methanol, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC20209CB
Specifications
| (4-Methyl-2-phenyl-5-pyrimidinyl)methanol | |
| 94.5 | |
| 95% | |
| C12H12N2O | |
| 250mg | |
| LXJHDOHHZSZIMJ-UHFFFAOYSA-N | |
| (4-methyl-2-phenylpyrimidin-5-yl)methanol | |
| 2776512 | |
| 95% |
| 342405-27-0 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD02682074 | |
| 4-methyl-2-phenylpyrimidin-5-yl methanol, 4-methyl-2-phenyl-5-pyrimidinyl methanol, 5-pyrimidinemethanol,4-methyl-2-phenyl, 4-methyl-2-phenylpyrimidin-5-yl methan-1-ol | |
| CC1=NC(=NC=C1CO)C2=CC=CC=C2 | |
| 200.241 | |
| 200.24 |
Chemical Identifiers
| 342405-27-0 | |
| 200.241 | |
| LXJHDOHHZSZIMJ-UHFFFAOYSA-N | |
| 2776512 | |
| CC1=NC(=NC=C1CO)C2=CC=CC=C2 |
| C12H12N2O | |
| MFCD02682074 | |
| 4-methyl-2-phenylpyrimidin-5-yl methanol, 4-methyl-2-phenyl-5-pyrimidinyl methanol, 5-pyrimidinemethanol,4-methyl-2-phenyl, 4-methyl-2-phenylpyrimidin-5-yl methan-1-ol | |
| (4-methyl-2-phenylpyrimidin-5-yl)methanol |