missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-2-phenyl-5-pyrimidinyl)methanol, 95%, Thermo Scientific™

Catalog No. p-2695140
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC20209CB 250mg
CC20209DA 1g
CC20209DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC20209CB Supplier Thermo Scientific Chemicals Supplier No. CC20209CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 342405-27-0
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.241
MDL Number MFCD02682074
InChI Key LXJHDOHHZSZIMJ-UHFFFAOYSA-N
Synonym 4-methyl-2-phenylpyrimidin-5-yl methanol, 4-methyl-2-phenyl-5-pyrimidinyl methanol, 5-pyrimidinemethanol,4-methyl-2-phenyl, 4-methyl-2-phenylpyrimidin-5-yl methan-1-ol
PubChem CID 2776512
IUPAC Name (4-methyl-2-phenylpyrimidin-5-yl)methanol
SMILES CC1=NC(=NC=C1CO)C2=CC=CC=C2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 200.24
Percent Purity 95%
Chemical Name or Material (4-Methyl-2-phenyl-5-pyrimidinyl)methanol