missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenyl-1,3-dioxolane (mixture of isomers) 98.0+%, TCI America™

Catalog No. B109425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B109425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B109425ML Supplier TCI America Supplier No. B109425ML
Only null left

Chemical Identifiers

CAS 2568-25-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00059732
InChI Key CDIKGISJRLTLRA-UHFFFAOYSA-N
Synonym benzaldehyde propylene glycol acetal, 1,3-dioxolane, 4-methyl-2-phenyl, 4-methyl-2-phenyl-m-dioxolane, benzaldehyde propylene glycolacetal, benzaldehyde propyleneglycol acetal, benzaldehyde propylene acetal, 4-methyl-2-phenyl-1.3-dioxolane, 4-methyl-2-phenyl-1,3 dioxolane, #, benzaldehydepropyleneglycolacetal
PubChem CID 595928
IUPAC Name 4-methyl-2-phenyl-1,3-dioxolane
SMILES CC1COC(O1)C2=CC=CC=C2

Specifications

Color Colorless
Boiling Point 125°C
Quantity 25 mL
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-phenyl-1,3-dioxolane (mixture of isomers)
EINECSNumber (5)-0518
RTECSNumber JI3870000
TSCA Yes