accessibility menu, dialog, popup

UserName

CAS RN 2568-25-4

CH3OO

CAS RN 2568-25-4

IUPAC Name: 4-methyl-2-phenyl-1,3-dioxolane
Synonyms: benzaldehyde propyleneglycol acetal
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: CDIKGISJRLTLRA-UHFFFAOYNA-N
SMILES: CC1COC(O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results