missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentyl Acetate 98.0+%, TCI America™

Catalog No. A134725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A134725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A134725G Supplier TCI America Supplier No. A134725G
Only null left

Chemical Identifiers

CAS 108-84-9
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00053723
InChI Key CPIVYSAVIPTCCX-UHFFFAOYNA-N
Synonym Acetic Acid 1,3-Dimethylbutyl Ester, Acetic Acid 4-Methyl-2-pentyl Ester, 1,3-Dimethylbutyl Acetate
PubChem CID 7959
IUPAC Name 4-methylpentan-2-yl acetate
SMILES CC(C)CC(C)OC(C)=O

Specifications

Color Colorless
Boiling Point 144°C
Quantity 25 g
UN Number 1233
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentyl Acetate
EINECSNumber (2)-0734
RTECSNumber SA7525000
TSCA Yes