CAS RN 108-84-9
CAS RN 108-84-9
IUPAC Name:
4-methylpentan-2-yl acetate
Synonyms:
4-methyl-2-pentyl acetate
Molecular Weight (g/mol):
144.21
Molecular Formula:
C8H16O2
InChi Key:
CPIVYSAVIPTCCX-UHFFFAOYNA-N
SMILES:
CC(C)CC(C)OC(C)=O
1
–
1
of
1
results
1
4-Methyl-2-pentyl Acetate 98.0+%, TCI Americaâ„¢
CAS: 108-84-9 Molecular Formula: C8H16O2 Molecular Weight (g/mol): 144.21 MDL Number: MFCD00053723 InChI Key: CPIVYSAVIPTCCX-UHFFFAOYNA-N Synonym: Acetic Acid 1,3-Dimethylbutyl Ester, Acetic Acid 4-Methyl-2-pentyl Ester, 1,3-Dimethylbutyl Acetate PubChem CID: 7959 IUPAC Name: 4-methylpentan-2-yl acetate SMILES: CC(C)CC(C)OC(C)=O