missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentene (cis- and trans- mixture) 96.0+%, TCI America™

Catalog No. p-7132141
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03951ML 1 mL
M03955ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M03951ML Supplier TCI America Supplier No. M03951ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4461-48-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00065138,MFCD00065138,MFCD00009293
InChI Key LGAQJENWWYGFSN-PLNGDYQASA-N
PubChem CID 5326159
IUPAC Name (2Z)-4-methylpent-2-ene
SMILES C\C=C/C(C)C

Specifications

Color Colorless
Boiling Point 58°C
Quantity 1 mL
UN Number 2288
Formula Weight 84.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentene (cis- and trans- mixture)
EINECSNumber (2)-0031
TSCA No