accessibility menu, dialog, popup

UserName

CH3CH3H3C(Z)

CAS RN 4461-48-7

IUPAC Name: (2Z)-4-methylpent-2-ene
Synonyms: (2Z)-4-methylpent-2-ene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: LGAQJENWWYGFSN-PLNGDYQASA-N
SMILES: C\C=C/C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results