missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanol 98.0+%, TCI America™

Catalog No. M038625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M038625ML 25 mL
M0386500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M038625ML Supplier TCI America Supplier No. M038625ML

Chemical Identifiers

CAS 108-11-2
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004550
InChI Key WVYWICLMDOOCFB-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanol, 2-pentanol, 4-methyl, isobutylmethylcarbinol, 2-methyl-4-pentanol, methyl amyl alcohol, methyl isobutyl carbinol, mibc, 1,3-dimethylbutanol, 4-methylpentanol-2, isobutylmethylmethanol
PubChem CID 7910
IUPAC Name 4-methylpentan-2-ol
SMILES CC(C)CC(C)O

Specifications

Melting Point -90°C
Color Colorless
Boiling Point 132°C
Quantity 25 mL
UN Number 2053
Formula Weight 102.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanol
EINECSNumber (2)-0217
RTECSNumber SA7350000
TSCA Yes