missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-oxovaleric Acid 95.0+%, TCI America™

Catalog No. K00255G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K00255G Supplier TCI America Supplier No. K00255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 816-66-0
Molecular Formula C6H9NaO3
Molecular Weight (g/mol) 152.13
MDL Number MFCD00066204
InChI Key IXFAZKRLPPMQEO-UHFFFAOYSA-M
Synonym 4-methyl-2-oxovaleric acid, alpha-ketoisocaproic acid, ketoleucine, alpha-ketoisocaproate, pentanoic acid, 4-methyl-2-oxo, 2-oxoisocaproate, 2-oxo-4-methylpentanoic acid, ketoisocaproate, 2-oxoisohexanoate, 4-methyl-2-oxopentanoate
PubChem CID 70
ChEBI CHEBI:48430
IUPAC Name sodium 4-methyl-2-oxopentanoate
SMILES [Na+].CC(C)CC(=O)C([O-])=O

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 85°C
Quantity 5 g
UN Number 3265
Formula Weight 130.14
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-oxovaleric Acid