accessibility menu, dialog, popup

UserName

NaH3CCH3OOO

CAS RN 816-66-0

IUPAC Name: sodium 4-methyl-2-oxopentanoate
Synonyms: α-ketoisocaproic acid
Molecular Weight (g/mol): 152.13
Molecular Formula: C6H9NaO3
InChi Key: IXFAZKRLPPMQEO-UHFFFAOYSA-M
SMILES: [Na+].CC(C)CC(=O)C([O-])=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)