Learn More
4-Methyl-2-oxovaleric acid, 94%
CAS: 816-66-0 | C6H9NaO3 | 152.13 g/mol
$128.70 - $453.68
Chemical Identifiers
| CAS | 816-66-0 |
|---|---|
| Molecular Formula | C6H9NaO3 |
| Molecular Weight (g/mol) | 152.13 |
| MDL Number | MFCD00066204 |
| InChI Key | IXFAZKRLPPMQEO-UHFFFAOYSA-M |
| Synonym | 4-methyl-2-oxovaleric acid, alpha-ketoisocaproic acid, ketoleucine, alpha-ketoisocaproate, pentanoic acid, 4-methyl-2-oxo, 2-oxoisocaproate, 2-oxo-4-methylpentanoic acid, ketoisocaproate, 2-oxoisohexanoate, 4-methyl-2-oxopentanoate |
| PubChem CID | 70 |
| ChEBI | CHEBI:48430 |
| SMILES | [Na+].CC(C)CC(=O)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6007603
|
Thermo Scientific Chemicals
H6007603 |
1 g |
Each for $128.70
|
|
|||||
|
AAH6007606
|
Thermo Scientific Chemicals
H6007606 |
5 g |
Each for $453.68
|
|
|||||
Description
Ii is an intermediate in the metabolism of leucine and also used as flavoring agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIi is an intermediate in the metabolism of leucine and also used as flavoring agents.
Solubility
Soluble in water.
Notes
Refrigerate in tightly sealed containers. Keep away from oxidizing agents.
Chemical Identifiers
| 816-66-0 | |
| 152.13 | |
| IXFAZKRLPPMQEO-UHFFFAOYSA-M | |
| 70 | |
| [Na+].CC(C)CC(=O)C([O-])=O |
| C6H9NaO3 | |
| MFCD00066204 | |
| 4-methyl-2-oxovaleric acid, alpha-ketoisocaproic acid, ketoleucine, alpha-ketoisocaproate, pentanoic acid, 4-methyl-2-oxo, 2-oxoisocaproate, 2-oxo-4-methylpentanoic acid, ketoisocaproate, 2-oxoisohexanoate, 4-methyl-2-oxopentanoate | |
| CHEBI:48430 |
Specifications
| 816-66-0 | |
| 1.055 | |
| C6H9NaO3 | |
| (CH3)2CHCH2COCOOH | |
| 1 g | |
| 1701823 | |
| Soluble in water. | |
| [Na+].CC(C)CC(=O)C([O-])=O | |
| 152.13 | |
| CHEBI:48430 | |
| 98% |
| 8°C to 10°C | |
| 82°C to 83°C | |
| 1.431 | |
| MFCD00066204 | |
| UN3265 | |
| 4-methyl-2-oxovaleric acid, alpha-ketoisocaproic acid, ketoleucine, alpha-ketoisocaproate, pentanoic acid, 4-methyl-2-oxo, 2-oxoisocaproate, 2-oxo-4-methylpentanoic acid, ketoisocaproate, 2-oxoisohexanoate, 4-methyl-2-oxopentanoate | |
| IXFAZKRLPPMQEO-UHFFFAOYSA-M | |
| sodium 4-methyl-2-oxopentanoate | |
| 70 | |
| 130.14 | |
| 4-Methyl-2-oxovaleric acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 212-435-5
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only