missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-oxovaleric acid, 94%

Catalog No. AAH6007603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6007603 1 g
AAH6007606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6007603 Supplier Thermo Scientific Chemicals Supplier No. H6007603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 816-66-0 | C6H9NaO3 | 152.13 g/mol

Ii is an intermediate in the metabolism of leucine and also used as flavoring agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ii is an intermediate in the metabolism of leucine and also used as flavoring agents.

Solubility
Soluble in water.

Notes
Refrigerate in tightly sealed containers. Keep away from oxidizing agents.

Chemical Identifiers

CAS 816-66-0
Molecular Formula C6H9NaO3
Molecular Weight (g/mol) 152.13
MDL Number MFCD00066204
InChI Key IXFAZKRLPPMQEO-UHFFFAOYSA-M
Synonym 4-methyl-2-oxovaleric acid, alpha-ketoisocaproic acid, ketoleucine, alpha-ketoisocaproate, pentanoic acid, 4-methyl-2-oxo, 2-oxoisocaproate, 2-oxo-4-methylpentanoic acid, ketoisocaproate, 2-oxoisohexanoate, 4-methyl-2-oxopentanoate
PubChem CID 70
ChEBI CHEBI:48430
SMILES [Na+].CC(C)CC(=O)C([O-])=O

Specifications

Melting Point 8°C to 10°C
Density 1.055
Boiling Point 82°C to 83°C
Refractive Index 1.431
Linear Formula (CH3)2CHCH2COCOOH
Quantity 1 g
UN Number UN3265
Beilstein 1701823
Solubility Information Soluble in water.
IUPAC Name sodium 4-methyl-2-oxopentanoate
Formula Weight 130.14
Percent Purity 98%
Chemical Name or Material 4-Methyl-2-oxovaleric acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-435-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only