missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-nitroaniline, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 112°C to 115°C |
| Color | Brown-Orange to Orange |
| Boiling Point | 169°C (21.0 mmHg) |
| Flash Point | 157°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | CH3C6H3(NO2)NH2 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Toxic if swallowed. Toxic if inhaled. Toxic to aquatic life with long lasting effects. Toxic in contact with skin. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 201-924-9 |
| RTECSNumber | XU8227250 |
| TSCA | TSCA |
RUO – Research Use Only