missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-nitroaniline, 98+%

Catalog No. AAA1366622
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1366622 100 g
AAA1366636 500 g
AAA136660E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1366622 Supplier Thermo Scientific Chemicals Supplier No. A1366622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 89-62-3 | C7H8N2O2 | 152.15 g/mol

4-Methyl-2-nitroaniline is used as a red azo dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-2-nitroaniline is used as a red azo dye.

Solubility
Soluble in water (0.2 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 89-62-3
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007907
InChI Key DLURHXYXQYMPLT-UHFFFAOYSA-N
Synonym 2-nitro-p-toluidine, 4-amino-3-nitrotoluene, 3-nitro-4-toluidine, fast red gl, 3-nitro-4-aminotoluene, fast red gl base, benzenamine, 4-methyl-2-nitro, 2-nitro-4-methylaniline, 4-methyl-6-nitroaniline, mnpt
PubChem CID 6978
ChEBI CHEBI:66920
IUPAC Name 4-methyl-2-nitroaniline
SMILES CC1=CC=C(N)C(=C1)[N+]([O-])=O

Specifications

Melting Point 115°C to 118°C
Density 1.31
Boiling Point 169°C (21 mmHg)
Flash Point 157°C (314°F)
Odor Characteristic
Quantity 100 g
UN Number UN2660
Beilstein 879506
Solubility Information Soluble in water (0.2g/L at 20°C).
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 4-Methyl-2-nitroaniline
Show More Show Less
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROTOLUIDINES (MONO)
EINECSNumber 201-924-9
RTECSNumber XU8227250
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only