missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2(5H)-furanone 97.0+%, TCI America™

Catalog No. M10795ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10795ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10795ML Supplier TCI America Supplier No. M10795ML

Chemical Identifiers

CAS 6124-79-4
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00191546
InChI Key ZZEYQBNQZKUWKY-UHFFFAOYSA-N
Synonym 4-Hydroxy-3-methyl-2-butenoic Acid gamma-Lactone
PubChem CID 145832
IUPAC Name 4-methyl-2,5-dihydrofuran-2-one
SMILES CC1=CC(=O)OC1

Specifications

Color Colorless
Boiling Point 113°C
Quantity 5 mL
Formula Weight 98.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-2(5H)-furanone