accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 6124-79-4

IUPAC Name: 4-methyl-2,5-dihydrofuran-2-one
Synonyms: 4-methyl-5H-furan-2-one
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: ZZEYQBNQZKUWKY-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results