missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 110.12 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Methyl-1H-imidazole-5-carbaldehyde |