missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-phenyl-3-pyrazolidone 98.0+%, TCI America™

Catalog No. M166725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M166725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M166725G Supplier TCI America Supplier No. M166725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2654-57-1
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.22
MDL Number MFCD00044800
InChI Key ZZEYCGJAYIHIAZ-UHFFFAOYNA-N
Synonym 1-phenyl-4-methyl-3-pyrazolidone, phenidone b, 4-methyl-1-phenyl-3-pyrazolidinone, 3-pyrazolidinone, 4-methyl-1-phenyl, 4-methyl-1-phenyl-3-pyrazolidone, 4-methyl-1-phenyl-pyrazolidin-3-one, 1-phenyl-4-methylpyrazolidone-3, 4-methyl-1-phenyl-4,5-dihydro-1h-pyrazol-3-ol, 4-methyl-1-phenyl-3-pyrazilidone, phenidone b 1-phenyl-4-methyl-3-pyrazolidone
PubChem CID 98282
IUPAC Name 4-methyl-1-phenylpyrazolidin-3-one
SMILES CC1CN(NC1=O)C1=CC=CC=C1

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 25 g
Formula Weight 176.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-1-phenyl-3-pyrazolidone
EINECSNumber (5)-3473
TSCA Yes