accessibility menu, dialog, popup

UserName

CAS RN 2654-57-1

IUPAC Name: 4-methyl-1-phenylpyrazolidin-3-one
Synonyms: 4-methyl-1-phenylpyrazolidin-3-one
Molecular Weight (g/mol): 176.22
Molecular Formula: C10H12N2O
InChi Key: ZZEYCGJAYIHIAZ-UHFFFAOYNA-N
SMILES: CC1CN(NC1=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results