missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-naphthylmagnesium bromide, 0.25M solution in THF, AcroSeal™, Thermo Scientific™

Catalog No. AC433690500
Change view
Click to view available options
Quantity:
50mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433690500 50mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433690500 Supplier Thermo Scientific Chemicals Supplier No. 433690500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 115913-58-1 , 109-99-9
Molecular Formula C11H9BrMg
Molecular Weight (g/mol) 245.4
InChI Key JJEPJVHQVFSQDI-UHFFFAOYSA-M
Synonym 4-methyl-1-naphthylmagnesium bromide, 4-methyl-1-naphthylmagnesium bromide, 0.25 m in thf, 4-methyl-1-naphthylmagnesium bromide, 0.25 m in 2-methf, 4-methyl-1-naphthylmagnesium bromide 0.25 m in tetrahydrofuran, grignard reagent
PubChem CID 13487699
IUPAC Name magnesium;4-methyl-1H-naphthalen-1-ide;bromide
SMILES CC1=CC=[C-]C2=CC=CC=C12.[Mg+2].[Br-]

Specifications

CAS Min % 5.7, 92.75
CAS Max % 7.25, 94.31
Density 0.9480g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Quantity 50mL
Specific Gravity 0.948
Solubility Information Solubility in water: reacts
Formula Weight 245.4
Chemical Name or Material 4-Methyl-1-naphthylmagnesium bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Suspected of causing cancer if inhaled.
May form explosive peroxides.
Reacts violently wit
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only