missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-naphthoic Acid 98.0+%, TCI America™

Catalog No. M12035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12035G 5 g
M120325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12035G Supplier TCI America Supplier No. M12035G
Only null left

Chemical Identifiers

CAS 4488-40-8
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00671557
InChI Key SIVYRLBDAPKADZ-UHFFFAOYSA-N
Synonym 4-methyl-1-naphthoic acid, 4-methyl-1-naphthalenecarboxylic acid, 4-methyl-1-napthoic acid, 1-naphthalenecarboxylic acid, 4-methyl, 4-methyl-naphthalene-1-carboxylic acid, 4-methylnaphthalenecarboxylic acid, 4-methylnaphthoic acid, acmc-1aehw, 4-methyl-1-naphthoicacid, 4-methyl-1-naphtholic acid
PubChem CID 3014285
IUPAC Name 4-methylnaphthalene-1-carboxylic acid
SMILES CC1=C2C=CC=CC2=C(C=C1)C(O)=O

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-1-naphthoic Acid