missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-acetonaphthone 97.0+%, TCI America™

Catalog No. p-7132596
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12025G 5 g
M120225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12025G Supplier TCI America Supplier No. M12025G
Only null left

Chemical Identifiers

CAS 28418-86-2
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00671556
InChI Key BKGIZYOJHJKFJP-UHFFFAOYSA-N
Synonym 1-Acetyl-4-methylnaphthalene
PubChem CID 97645
IUPAC Name 1-(4-methylnaphthalen-1-yl)ethanone
SMILES CC1=CC=C(C2=CC=CC=C12)C(=O)C

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 184.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methyl-1-acetonaphthone