missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-thiazole-5-sulfonyl chloride, 97%

Catalog No. p-7077426
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453640010 1 g
AC453640050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453640010 Supplier Thermo Scientific Chemicals Supplier No. 453640010
Only null left

CAS: 953070-51-4 | C4H4ClNO2S2 | 197.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 953070-51-4
Molecular Formula C4H4ClNO2S2
Molecular Weight (g/mol) 197.66
InChI Key AWTJUYXVSWZCIK-UHFFFAOYSA-N
Synonym 4-methylthiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 4-methyl, chloro 4-methyl 1,3-thiazol-5-yl sulfone
PubChem CID 18537519
IUPAC Name 4-methyl-1,3-thiazole-5-sulfonyl chloride
SMILES CC1=C(SC=N1)S(=O)(=O)Cl

Specifications

Melting Point 34.0°C to 36.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 197.66
Percent Purity 97%
Chemical Name or Material 4-Methyl-1,3-thiazole-5-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

RUO – Research Use Only