missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-dioxane 99.0+%, TCI America™

Catalog No. M020925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M020925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M020925ML Supplier TCI America Supplier No. M020925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1120-97-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00006569
InChI Key INCCMBMMWVKEGJ-UHFFFAOYSA-N
Synonym 4-methyldioxane, 4-methyl-m-dioxane, 1,3-dioxane, 4-methyl, m-dioxane, 4-methyl, 1,3-butanediol formal, 4-methyl-1,3 dioxane, acmc-2099cl, 5-19-01-00054 beilstein handbook reference, 4-methyl-1,3-dioxan-2-yl, m-dioxane, 4-methyl-8ci
PubChem CID 14269
IUPAC Name 4-methyl-1,3-dioxane
SMILES CC1CCOCO1

Specifications

Color Colorless
Boiling Point 114°C
Quantity 25 mL
UN Number 1993
Formula Weight 102.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-1,3-dioxane
EINECSNumber (5)-0838
RTECSNumber JH2200000
TSCA No