accessibility menu, dialog, popup

UserName

CAS RN 1120-97-4

IUPAC Name: 4-methyl-1,3-dioxane
Synonyms: 4-methyldioxane
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: INCCMBMMWVKEGJ-UHFFFAOYNA-N
SMILES: CC1CCOCO1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)