Learn More
4-Methoxytrityl chloride, 97%
CAS: 14470-28-1 | C20H17ClO | 308.805 g/mol
Supplier: Thermo Scientific Chemicals A1054518
Description
It is used as selective protecting reagent that is used for primary hydroxyl groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as selective protecting reagent that is used for primary hydroxyl groups.
Solubility
Soluble in toluene.
Notes
It is hygroscopic. Incompatible with oxidizing agents.
Specifications
| 4-Methoxytrityl chloride | |
| 121°C to 125°C | |
| MFCD00000814 | |
| 798425 | |
| 4-methoxytriphenylchloromethane, 4-methoxytrityl chloride, 4-methoxytriphenylmethyl chloride, 4-monomethoxytrityl chloride, mmtrcl, p-chlorodiphenylmethyl anisole, p-anisylchlorodiphenylmethane, 1-chlorodiphenylmethyl-4-methoxybenzene, chloro 4-methoxyphenyl methylene dibenzene, 4-methoxytritylchloride | |
| OBOHMJWDFPBPKD-UHFFFAOYSA-N | |
| 1-[chloro(diphenyl)methyl]-4-methoxybenzene | |
| 84462 | |
| 97% |
| 14470-28-1 | |
| C20H17ClO | |
| 50 g | |
| Moisture sensitive | |
| Soluble in toluene. | |
| COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl | |
| 308.805 | |
| 308.81 |
Chemical Identifiers
| 14470-28-1 | |
| 308.805 | |
| OBOHMJWDFPBPKD-UHFFFAOYSA-N | |
| 84462 | |
| COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl |
| C20H17ClO | |
| MFCD00000814 | |
| 4-methoxytriphenylchloromethane, 4-methoxytrityl chloride, 4-methoxytriphenylmethyl chloride, 4-monomethoxytrityl chloride, mmtrcl, p-chlorodiphenylmethyl anisole, p-anisylchlorodiphenylmethane, 1-chlorodiphenylmethyl-4-methoxybenzene, chloro 4-methoxyphenyl methylene dibenzene, 4-methoxytritylchloride | |
| 1-[chloro(diphenyl)methyl]-4-methoxybenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-463-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only