missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxytrityl alcohol, 94%

Catalog No. AAA1862009
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1862009 10 g
AAA1862018 50 g
AAA1862030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1862009 Supplier Thermo Scientific Chemicals Supplier No. A1862009

CAS: 847-83-6 | C20H18O2 | 290.36 g/mol

It is used as an intermediate for preparation of TT-0011.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate for preparation of TT-0011.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 847-83-6
Molecular Formula C20H18O2
Molecular Weight (g/mol) 290.36
MDL Number MFCD00087962
InChI Key WCRRRAKYYPJJMP-UHFFFAOYSA-N
Synonym p-methoxytrityl alcohol, 4-methoxyphenyl diphenylmethanol, unii-d117s0gbot, 4-methoxytrityl alcohol, p-methoxyphenyl-diphenylmethanol, p-anisyldiphenylmethanol, d117s0gbot, 4-methoxyphenyldiphenylmethanol, methoxytriphenylcarbinole
PubChem CID 70061
IUPAC Name (4-methoxyphenyl)-diphenylmethanol
SMILES COC1=CC=C(C=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 290.36
Percent Purity 94%
Chemical Name or Material 4-Methoxytrityl alcohol
EINECSNumber 212-693-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only