missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxythiophenol, 97%

Catalog No. aaa1600906
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAA1600906 5 g
AAA1600914 25 g
AAA1600922 100 g
3 options

Catalog No. AAA1600906

Supplier: Thermo Scientific Chemicals A1600906

Only null left

CAS: 696-63-9 | C7H8OS | 140.2 g/mol

4-Methoxythiophenol is used in preparing samples for self-assembled monolayers (SAM?s) characterization and to study SAM’s effect on both n- and p-channel organic thin film transistors. It is also used in cesium fluoride-Celite catalyzed preparation of thioethers and thioesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxythiophenol is used in preparing samples for self-assembled monolayers (SAM′s) characterization and to study SAM’s effect on both n- and p-channel organic thin film transistors. It is also used in cesium fluoride-Celite catalyzed preparation of thioethers and thioesters.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.

Chemical Identifiers

CAS 696-63-9
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00004849
InChI Key NIFAOMSJMGEFTQ-UHFFFAOYSA-N
Synonym 4-methoxythiophenol, p-methoxybenzenethiol, benzenethiol, 4-methoxy, 4-methoxybenzene-1-thiol, benzenethiol, p-methoxy, 4-mercaptoanisole, 4-methoxy thiophenol, 4-methoxy-benzenethiol, para-methoxybenzenethiol, 4-mercaptoanisol
PubChem CID 12765
IUPAC Name 4-methoxybenzenethiol
SMILES COC1=CC=C(C=C1)S

Specifications

Density 1.14
Boiling Point 100°C to 103°C (13 mmHg)
Flash Point 96°C (204°F)
Odor Unpleasant
Refractive Index 1.5831
Quantity 5 g
Beilstein 1446910
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix.
Formula Weight 140.21
Percent Purity 97%
Chemical Name or Material 4-Methoxythiophenol
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 211-799-2
RTECSNumber DC1800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only