missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxythiobenzamide, 98%

Catalog No. AAL1094506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1094506 5 g
AAL1094503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1094506 Supplier Thermo Scientific Chemicals Supplier No. L1094506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2362-64-3 | C8H9NOS | 167.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2362-64-3
Molecular Formula C8H9NOS
Molecular Weight (g/mol) 167.226
MDL Number MFCD00040993
InChI Key WKWVTPKUHJOVTI-UHFFFAOYSA-N
Synonym 4-methoxythiobenzamide, 4-methoxybenzothioamide, 4-methoxybenzene-1-carbothioamide, benzenecarbothioamide, 4-methoxy, 4-methoxy-thiobenzamide, p-methoxythiobenzamide, amino 4-methoxyphenyl methane-1-thione, thio-p-anisamide, p-anisamide,thio, p-methoxybenzothiamide
PubChem CID 736827
IUPAC Name 4-methoxybenzenecarbothioamide
SMILES COC1=CC=C(C=C1)C(=S)N

Specifications

Melting Point 145°C to 147°C
Quantity 5 g
Beilstein 1072444
Formula Weight 167.23
Percent Purity 98%
Chemical Name or Material 4-Methoxythiobenzamide
Hazard Category H302+H332
Hazard Statement GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only