missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxythioanisole 98.0+%, TCI America™
$63.38 - $268.43
Chemical Identifiers
| CAS | 1879-16-9 |
|---|---|
| Molecular Formula | C8H10OS |
| Molecular Weight (g/mol) | 154.227 |
| MDL Number | MFCD00010587 |
| InChI Key | DQNSKXYRRRCKGH-UHFFFAOYSA-N |
| Synonym | 4-methoxythioanisole, 1-methoxy-4-methylthio benzene, 4-methylthio anisole, p-methylthio anisole, p-anisyl methyl sulfide, p-methoxyphenyl methyl sulfide, 1-methoxy-4-methylthiobenzene, benzene, 1-methoxy-4-methylthio, anisole, p-methylthio, 4-methoxy thioanisole |
| PubChem CID | 15885 |
| IUPAC Name | 1-methoxy-4-methylsulfanylbenzene |
| SMILES | COC1=CC=C(C=C1)SC |
Chemical Identifiers
| 1879-16-9 | |
| 154.227 | |
| DQNSKXYRRRCKGH-UHFFFAOYSA-N | |
| 15885 | |
| COC1=CC=C(C=C1)SC |
| C8H10OS | |
| MFCD00010587 | |
| 4-methoxythioanisole, 1-methoxy-4-methylthio benzene, 4-methylthio anisole, p-methylthio anisole, p-anisyl methyl sulfide, p-methoxyphenyl methyl sulfide, 1-methoxy-4-methylthiobenzene, benzene, 1-methoxy-4-methylthio, anisole, p-methylthio, 4-methoxy thioanisole | |
| 1-methoxy-4-methylsulfanylbenzene |
Specifications
| 1879-16-9 | |
| 137°C | |
| MFCD00010587 | |
| 4-methoxythioanisole, 1-methoxy-4-methylthio benzene, 4-methylthio anisole, p-methylthio anisole, p-anisyl methyl sulfide, p-methoxyphenyl methyl sulfide, 1-methoxy-4-methylthiobenzene, benzene, 1-methoxy-4-methylthio, anisole, p-methylthio, 4-methoxy thioanisole | |
| COC1=CC=C(C=C1)SC | |
| 154.227 | |
| 154.23 | |
| Liquid |
| Yellow | |
| C8H10OS | |
| 5 g | |
| DQNSKXYRRRCKGH-UHFFFAOYSA-N | |
| 1-methoxy-4-methylsulfanylbenzene | |
| 15885 | |
| ≥98.0% (GC) | |
| 4-Methoxythioanisole |
Safety and Handling
TSCA : No