missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxystyrene (stabilized with TBC) 98.0+%, TCI America™

Catalog No. M013025ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M013025ML 25 mL
M013010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M013025ML Supplier TCI America Supplier No. M013025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 637-69-4
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008619
InChI Key UAJRSHJHFRVGMG-UHFFFAOYSA-N
Synonym 4-methoxystyrene, 1-methoxy-4-vinylbenzene, p-methoxystyrene, 4-vinylanisole, benzene, 1-ethenyl-4-methoxy, p-vinylanisole, anisole, p-vinyl, unii-2ish8t4a6e, ccris 4381, 4-vinylanisol
PubChem CID 12507
IUPAC Name 1-ethenyl-4-methoxybenzene
SMILES COC1=CC=C(C=C)C=C1

Specifications

Boiling Point 98°C
Quantity 25 mL
Formula Weight 134.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxystyrene (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator