missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxystyrene, 96%, stabilized

Catalog No. AC161390250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161390250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC161390250 Supplier Thermo Scientific Chemicals Supplier No. 161390250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 637-69-4 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 637-69-4
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008619
InChI Key UAJRSHJHFRVGMG-UHFFFAOYSA-N
Synonym 4-methoxystyrene, 1-methoxy-4-vinylbenzene, p-methoxystyrene, 4-vinylanisole, benzene, 1-ethenyl-4-methoxy, p-vinylanisole, anisole, p-vinyl, unii-2ish8t4a6e, ccris 4381, 4-vinylanisol
PubChem CID 12507
IUPAC Name 1-ethenyl-4-methoxybenzene
SMILES COC1=CC=C(C=C)C=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 204°C to 205°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5600 to 1.562
Linear Formula H2C=CHC6H4OCH3
Quantity 25 g
Beilstein 06, 561
Specific Gravity 1
Stabilizer 0.1% 4-tert-butylcatechol
Formula Weight 134.18
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Methoxystyrene, 96%
Show More Show Less
EINECSNumber 211-298-9

RUO – Research Use Only