missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxysalicylic Acid 98.0+%, TCI America™

Catalog No. p-7132856
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17955G 5 g
M179525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17955G Supplier TCI America Supplier No. M17955G
Only null left

Chemical Identifiers

CAS 2237-36-7
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002450
InChI Key MRIXVKKOHPQOFK-UHFFFAOYSA-N
Synonym 4-methoxysalicylic acid, 2-hydroxy-p-anisic acid, 2-hydroxy-4-methoxy-benzoic acid, benzoic acid, 2-hydroxy-4-methoxy, 2-hydroxy-4-methoxybenzoes, 4-methoxysalicylicacid, pubchem18895, 4-methoxy salicylic acid, 4-methoxy-salicylic acid, acmc-1cs6b
PubChem CID 75231
IUPAC Name 2-hydroxy-4-methoxybenzoic acid
SMILES COC1=CC=C(C(O)=O)C(O)=C1

Specifications

Melting Point 160°C
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxysalicylic Acid