missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxysalicylic acid, 99%
CAS: 2237-36-7 | C8H8O4 | 168.15 g/mol
$189.85 - $189.85
Chemical Identifiers
| CAS | 2237-36-7 |
|---|---|
| Molecular Formula | C8H8O4 |
| Molecular Weight (g/mol) | 168.15 |
| MDL Number | MFCD00002450 |
| InChI Key | MRIXVKKOHPQOFK-UHFFFAOYSA-N |
| Synonym | 4-methoxysalicylic acid, 2-hydroxy-p-anisic acid, 2-hydroxy-4-methoxy-benzoic acid, benzoic acid, 2-hydroxy-4-methoxy, 2-hydroxy-4-methoxybenzoes, 4-methoxysalicylicacid, pubchem18895, 4-methoxy salicylic acid, 4-methoxy-salicylic acid, acmc-1cs6b |
| PubChem CID | 75231 |
| IUPAC Name | 2-hydroxy-4-methoxybenzoic acid |
| SMILES | COC1=CC=C(C(O)=O)C(O)=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC174240100
|
Thermo Scientific Chemicals
174240100 |
10 g | Glass bottle |
Each for $189.85
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2237-36-7 | |
| 168.15 | |
| MRIXVKKOHPQOFK-UHFFFAOYSA-N | |
| 75231 | |
| COC1=CC=C(C(O)=O)C(O)=C1 |
| C8H8O4 | |
| MFCD00002450 | |
| 4-methoxysalicylic acid, 2-hydroxy-p-anisic acid, 2-hydroxy-4-methoxy-benzoic acid, benzoic acid, 2-hydroxy-4-methoxy, 2-hydroxy-4-methoxybenzoes, 4-methoxysalicylicacid, pubchem18895, 4-methoxy salicylic acid, 4-methoxy-salicylic acid, acmc-1cs6b | |
| 2-hydroxy-4-methoxybenzoic acid |
Specifications
| 2237-36-7 | |
| 100.0 | |
| White to Beige | |
| 98.5% min. (HPLC) | |
| C8H8O4 | |
| MFCD00002450 | |
| 10, 379 | |
| MRIXVKKOHPQOFK-UHFFFAOYSA-N | |
| 2-hydroxy-4-methoxybenzoic acid | |
| 75231 | |
| 99% | |
| 4-Methoxysalicylic acid, 99% |
| 98.5 | |
| 157.0°C to 159.0°C | |
| Authentic | |
| Glass bottle | |
| CH3OC6H3(OH)CO2H | |
| 10 g | |
| 4-methoxysalicylic acid, 2-hydroxy-p-anisic acid, 2-hydroxy-4-methoxy-benzoic acid, benzoic acid, 2-hydroxy-4-methoxy, 2-hydroxy-4-methoxybenzoes, 4-methoxysalicylicacid, pubchem18895, 4-methoxy salicylic acid, 4-methoxy-salicylic acid, acmc-1cs6b | |
| COC1=CC=C(C(O)=O)C(O)=C1 | |
| 168.15 | |
| 168.15 | |
| Powder |
Safety and Handling
EINECSNumber : 218-801-0
RUO – Research Use Only