missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxypropiophenone, 99%

Catalog No. AAA1461914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1461914 25 g
AAA1461922 100 g
AAA1461936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1461914 Supplier Thermo Scientific Chemicals Supplier No. A1461914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 121-97-1 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-97-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00009310
InChI Key ZJVAWPKTWVFKHG-UHFFFAOYSA-N
PubChem CID 67144
IUPAC Name 1-(4-methoxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)OC

Specifications

Melting Point 27°C to 29°C
Density 1.07
Boiling Point 273°C to 275°C
Flash Point 62°C (143°F)
Refractive Index 1.5465
Quantity 25 g
Beilstein 907733
Synonym 4'-methoxypropiophenone, p-methoxypropiophenone, 4-methoxypropiophenone, 1-4-methoxyphenyl propan-1-one, 1-propanone, 1-4-methoxyphenyl, propiophenone, 4'-methoxy, ethyl 4-methoxyphenyl ketone, 1-4-methoxyphenyl-1-propanone, 1-propanone,1-4-methoxyphenyl, 1-4-methoxy-phenyl-propan-1-one
Formula Weight 164.2
Percent Purity 99%
Chemical Name or Material 4'-Methoxypropiophenone
Show More Show Less
Hazard Category H227-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 204-512-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only