missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. m063725g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
M063725G 25 g
M06375G 5 g
2 options

Catalog No. M063725G

Supplier: TCI America M063725G

Only null left

Chemical Identifiers

CAS 19501-58-7
Molecular Formula C7H11ClN2O
Molecular Weight (g/mol) 174.628
MDL Number MFCD00012945
InChI Key FQHCPFMTXFJZJS-UHFFFAOYSA-N
Synonym 4-methoxyphenylhydrazine hydrochloride, 4-methoxyphenyl hydrazine hydrochloride, 4-anisylhydrazine hydrochloride, 4-methoxyphenylhydrazine hcl, p-methoxyphenylhydrazine hydrochloride, 4-methoxyphenylhydrazine.hcl, 1-4-methoxyphenyl hydrazine hydrochloride, p-anisylhydrazine hydrochloride, 4-hydrazinoanisole hydrochloride, hydrazine, 4-methoxyphenyl-, monohydrochloride
PubChem CID 2723904
IUPAC Name (4-methoxyphenyl)hydrazine;hydrochloride
SMILES COC1=CC=C(C=C1)NN.Cl

Specifications

Color Purple-White
Quantity 25 g
Formula Weight 174.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenylhydrazine Hydrochloride