missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$134.75 - $134.75
Chemical Identifiers
| CAS | 5720-07-0 |
|---|---|
| Molecular Formula | C7H9BO3 |
| MDL Number | MFCD00039139 |
| InChI Key | VOAAEKKFGLPLLU-UHFFFAOYSA-N |
| PubChem CID | 201262 |
| IUPAC Name | (4-methoxyphenyl)boronic acid |
Chemical Identifiers
| 5720-07-0 | |
| MFCD00039139 | |
| 201262 |
| C7H9BO3 | |
| VOAAEKKFGLPLLU-UHFFFAOYSA-N | |
| (4-methoxyphenyl)boronic acid |
Specifications
| 5720-07-0 | |
| MFCD00039139 | |
| 4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacid | |
| COC1=CC=C(C=C1)B(O)O | |
| 151.96 | |
| 151.96 | |
| 4-Methoxyphenylboronic Acid (contains varying amounts of Anhydride) |
| C7H9BO3 | |
| 5 g | |
| VOAAEKKFGLPLLU-UHFFFAOYSA-N | |
| (4-methoxyphenyl)boronic acid | |
| 201262 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CY8975000
TSCA : No