accessibility menu, dialog, popup

UserName

H3COBOHHO

CAS RN 5720-07-0

IUPAC Name: (4-methoxyphenyl)boronic acid
Synonyms: 4-methoxyphenylboronic acid
Molecular Weight (g/mol): 151.96
Molecular Formula: C7H9BO3
InChi Key: VOAAEKKFGLPLLU-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)B(O)O
Quantity 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (4)