missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M12525G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12525G Supplier TCI America Supplier No. M12525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5720-07-0
Molecular Formula C7H9BO3
MDL Number MFCD00039139
InChI Key VOAAEKKFGLPLLU-UHFFFAOYSA-N
PubChem CID 201262
IUPAC Name (4-methoxyphenyl)boronic acid

Specifications

Quantity 5 g
Synonym 4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacid
SMILES COC1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 151.96
Formula Weight 151.96
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenylboronic Acid (contains varying amounts of Anhydride)
RTECSNumber CY8975000
TSCA No