missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylboronic acid, 97%

Catalog No. AC309480010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC309480010 1 g Glass bottle
AC309480050 5 g Glass bottle
AC309480250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC309480010 Supplier Thermo Scientific Chemicals Supplier No. 309480010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5720-07-0 | C7H9BO3 | 151.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5720-07-0
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD00039139
InChI Key VOAAEKKFGLPLLU-UHFFFAOYSA-N
Synonym 4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacid
PubChem CID 201262
IUPAC Name (4-methoxyphenyl)boronic acid
SMILES COC1=CC=C(C=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 208.0°C to 210.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H4B(OH)2
Quantity 1 g
Formula Weight 151.96
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue

RUO – Research Use Only