missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetylene, 98%

Catalog No. AAH2628303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2628303 1 g
AAH2628306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2628303 Supplier Thermo Scientific Chemicals Supplier No. H2628303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 768-60-5 | C9H8O | 132.162 g/mol

4-Ethynylanisole was used in the synthesis of photo luminescent 1,2-dihydrophosphinines via a [4 + 2] cycloaddition. Along with an arylboronic acid and sodium azide in a copper-catalyzed, three-component synthesis of trisubstituted 1,2,4-triazoles. In a study of a gold (III)-catalyzed hydroamination of alkynes leading to N-vinylindoles3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethynylanisole was used in the synthesis of photo luminescent 1,2-dihydrophosphinines via a [4 + 2] cycloaddition. Along with an arylboronic acid and sodium azide in a copper-catalyzed, three-component synthesis of trisubstituted 1,2,4-triazoles. In a study of a gold (III)-catalyzed hydroamination of alkynes leading to N-vinylindoles3.

Solubility
Insoluble in water. Soluble in chloroform, acetone, dichloromethane, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 768-60-5
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00168815
InChI Key KBIAVTUACPKPFJ-UHFFFAOYSA-N
Synonym 4-ethynylanisole, 4-methoxyphenylacetylene, 4'-methoxyphenyl acetylene, 1-ethynyl-4-methoxy-benzene, 1-eth-1-ynyl-4-methoxybenzene, benzene, 1-ethynyl-4-methoxy, 4-methoxyphenyl acetylene, 4-ethynyl-1-methoxybenzene, 4'-methoxyphenylacetylene, p-ethynylanisole
PubChem CID 251020
IUPAC Name 1-ethynyl-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C#C

Specifications

Melting Point 27°C to 29°C
Density 1.019
Boiling Point 87°C to 91°C (11 mmHg)
Flash Point 82°C (180°F)
Refractive Index 1.563
Quantity 1 g
Solubility Information Insoluble in water. Soluble in chloroform,acetone,dichloromethane,and methanol.
Formula Weight 132.16
Percent Purity 99%
Chemical Name or Material 4-Methoxyphenylacetylene
Hazard Category H315-H319-H335-H500
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only