missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetonitrile 97.0+%, TCI America™

Catalog No. M085625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M085625G 25 g
M0856500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M085625G Supplier TCI America Supplier No. M085625G

Chemical Identifiers

CAS 104-47-2
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00001919
InChI Key PACGLQCRGWFBJH-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetonitrile, 2-4-methoxyphenyl acetonitrile, 4-methoxybenzyl cyanide, 4-methoxyphenyl acetonitrile, p-methoxyphenylacetonitrile, benzeneacetonitrile, 4-methoxy, p-methoxybenzyl cyanide, anisylacetonitrile, 4-anisyl cyanide, p-methoxybenzeneacetonitrile
PubChem CID 66031
IUPAC Name 2-(4-methoxyphenyl)acetonitrile
SMILES COC1=CC=C(C=C1)CC#N

Specifications

Color Yellow
Boiling Point 290°C
Quantity 25 g
UN Number 3276
Formula Weight 147.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenylacetonitrile
RTECSNumber AM0810000
TSCA Yes