missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetic acid, 99%

Catalog No. p-158641
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126020050 5 g Glass bottle
AC126021000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC126020050 Supplier Thermo Scientific Chemicals Supplier No. 126020050

CAS: 104-01-8 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-01-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004345
InChI Key NRPFNQUDKRYCNX-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetic acid, homoanisic acid, 2-4-methoxyphenyl acetic acid, 4-methoxybenzeneacetic acid, benzeneacetic acid, 4-methoxy, 4-methoxyphenyl acetic acid, p-methoxyphenylacetic acid, 2-p-anisyl acetic acid, p-methoxyphenyl acetic acid
PubChem CID 7690
ChEBI CHEBI:55501
IUPAC Name 2-(4-methoxyphenyl)acetic acid
SMILES COC1=CC=C(C=C1)CC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 84.0°C to 88.0°C
Color White to Yellow
Boiling Point 140.0°C (3.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CO2H
Quantity 5 g
Beilstein 10, 190
Solubility Information Solubility in water: 6g/L (20 c)
Formula Weight 166.18
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Methoxyphenylacetic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Wear eye protection/face protection.
EINECSNumber 203-166-4
RTECSNumber AI8960000
TSCA TSCA

RUO – Research Use Only