missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetic acid, 98+%

Catalog No. AAA1439936
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1439936 500 g
AAA1439906 5 g
AAA1439922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1439936 Supplier Thermo Scientific Chemicals Supplier No. A1439936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104-01-8 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-01-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00004345
InChI Key NRPFNQUDKRYCNX-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetic acid, homoanisic acid, 2-4-methoxyphenyl acetic acid, 4-methoxybenzeneacetic acid, benzeneacetic acid, 4-methoxy, 4-methoxyphenyl acetic acid, p-methoxyphenylacetic acid, 2-p-anisyl acetic acid, p-methoxyphenyl acetic acid
PubChem CID 7690
ChEBI CHEBI:55501
IUPAC Name 2-(4-methoxyphenyl)acetic acid
SMILES COC1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 84°C to 88°C
Boiling Point 138°C to 140°C (3 mmHg)
Quantity 500 g
Beilstein 1101737
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 4-Methoxyphenylacetic acid
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 203-166-4
RTECSNumber AI8960000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only