missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Methoxyphenyl)urea 98.0+%, TCI America™
Supplier: TCI America M25755G
Specifications
| (4-Methoxyphenyl)urea | |
| 162°C | |
| C8H10N2O2 | |
| 5 g | |
| PGUKYDVWVXRPKK-UHFFFAOYSA-N | |
| (4-methoxyphenyl)urea | |
| 74066 | |
| ≥98.0% (HPLC,N) |
| 1566-42-3 | |
| White | |
| MFCD00014789 | |
| 4-methoxyphenyl urea, urea, 4-methoxyphenyl, p-anisylurea, 1-4-methoxyphenyl urea, urea, p-methoxyphenyl, p-methoxyphenyl urea, p-methoxyphenylurea, n-4-methoxyphenyl urea, amino-n-4-methoxyphenyl amide, acmc-20amej | |
| COC1=CC=C(NC(N)=O)C=C1 | |
| 166.18 | |
| 166.18 | |
| Crystalline Powder |
Chemical Identifiers
| 1566-42-3 | |
| 166.18 | |
| PGUKYDVWVXRPKK-UHFFFAOYSA-N | |
| 74066 | |
| COC1=CC=C(NC(N)=O)C=C1 |
| C8H10N2O2 | |
| MFCD00014789 | |
| 4-methoxyphenyl urea, urea, 4-methoxyphenyl, p-anisylurea, 1-4-methoxyphenyl urea, urea, p-methoxyphenyl, p-methoxyphenyl urea, p-methoxyphenylurea, n-4-methoxyphenyl urea, amino-n-4-methoxyphenyl amide, acmc-20amej | |
| (4-methoxyphenyl)urea |
Safety and Handling
RTECSNumber : YT6451200
TSCA : No